Search
Search

LOC100360417| Gene Symbol: LOC100360417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761232.1 | XP_008759454.1 | 8 - 9 | 2415 | 88 | |
| XM_008761233.1 | XP_008759455.1 | 8 - 9 | 2615 | 88 | ||
| XM_008761234.1 | XP_008759456.1 | 7 - 8 | 2004 | 88 | ||
| XM_008761235.1 | XP_008759457.1 | 6 - 7 | 549 | 88 | ||
| XM_008761236.1 | XP_008759458.1 | 6 - 7 | 1296 | 88 | ||
| XM_008761237.1 | XP_008759459.1 | 6 - 7 | 680 | 88 | ||
Set Membership: |
|