Search
Search
| Gene Symbol: Il6r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017020.3 | NP_058716.2 | 8 - 9 | 1070 | 103 | |
| XM_006232587.2 | XP_006232649.1 | 8 - 9 | 2504 | 103 | ||
| XM_006232588.2 | XP_006232650.1 | 8 - 9 | 2503 | 103 | ||
| XM_008761127.1 | XP_008759349.1 | 8 - 9 | 2496 | 103 | ||
| GenBank mRNA | M58587.1 | - | 8 - 9 | 1159 | 103 | |
Set Membership: |
|