Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01496390_m1
          See other PSMD12 GE Assays ›
          Gene Symbol:
          Psmd12
          Gene Name:
          proteasome (prosome, macropain) 26S subunit, non-ATPase, 12
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 95706808 - 95725452 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01496390_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmd12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005875.1 NP_001005875.1 10 - 11 1209 130
          GenBank mRNA BC083758.1 - 10 - 11 1209 7192849 130
          FQ218292.1 - 4 - 5 436 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001823.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmd12

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s)

          proteasome-mediated ubiquitin-dependent protein catabolic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P504124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162831 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8268 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106410 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P504124 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P504124 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P504124 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P546459 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74091 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24466 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10739226 - -
          Affymetrix Rat Gene 1.0 ST Array 10739227 - -
          Affymetrix Rat Gene 1.0 ST Array 10739228 - -
          Affymetrix Rat Gene 1.0 ST Array 10739231 - -
          Affymetrix Rat Gene 1.0 ST Array 10739230 - -
          Affymetrix Rat Gene 1.0 ST Array 10739229 - -
          Affymetrix Rat Gene 1.0 ST Array 10739232 - -
          Affymetrix Rat Gene 1.0 ST Array 10739233 - -
          Affymetrix Rat Gene 1.0 ST Array 10739234 - -
          Affymetrix Rat Gene 1.0 ST Array 10739235 - -
          Affymetrix Rat Gene 1.1 ST Array 10739225 - -
          Rat Genome 230 2.0 Array 1392983_PM_at - -
          Rat Genome 230 2.0 Array 1392983_at - -
          Affymetrix HT Rat Focus Array 1392983_at - -
          Affymetrix HT Rat Focus Array 1392983_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000016548.rn.1 TC1000001823.rn.1 RTA TC
          Rat Expression Set 230 1392983_PM_at - -
          Rat Expression Set 230 1392983_at - -
          Affymetrix Rat Gene 1.0 ST Array 10739225 - -
          Rat Genome U34 Set rc_AI044322_at - -
          Rat Genome U34 Set rc_AA957682_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20722564 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356243 - -
          RatRef-12 Expression BeadChip ILMN_57012 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline