Search
Search

Psmc3ip| Gene Symbol: Psmc3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134458.2 | NP_604453.2 | 5 - 6 | 495 | 82 | |
| XM_006247249.2 | XP_006247311.1 | 5 - 6 | 1347 | 82 | ||
| GenBank mRNA | AF352812.1 | - | 5 - 6 | 495 | 82 | |
| CK840952.1 | - | 3 - 4 | 82 | |||
Set Membership: |
|