Search
Search
| Gene Symbol: Aipl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021590.1 | NP_067601.1 | 1 - 2 | 160 | 83 | |
| XM_006246798.2 | XP_006246860.1 | 1 - 2 | 192 | 83 | ||
| GenBank mRNA | AF180340.1 | - | 1 - 2 | 160 | 83 | |
Set Membership: |
|