Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01497143_g1
See other STAT5A GE Assays ›
Gene Symbol:
Stat5a
Gene Name:
signal transducer and activator of transcription 5A
Gene Aliases:
Stat5
Chromosome Location:
Chr.10: 88764732 - 88789060 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01497143_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Stat5a
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_017064.1
NP_058760.1
15 - 16
1958
116
GenBank mRNA
U24175.1
-
15 - 16
1958
116
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1000001639.rn.1
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Apoptosis
Detect Genomic DNA
Rattus norvegicus
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Stat5a
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
allantoin metabolic process
luteinization
natural killer cell differentiation
citrate metabolic process
2-oxoglutarate metabolic process
succinate metabolic process
oxaloacetate metabolic process
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
isoleucine metabolic process
valine metabolic process
creatine metabolic process
fatty acid metabolic process
signal transduction
JAK-STAT cascade
female pregnancy
lactation
positive regulation of cell proliferation
peptidyl-tyrosine phosphorylation
regulation of steroid metabolic process
cytokine-mediated signaling pathway
taurine metabolic process
lipid storage
regulation of cell adhesion
regulation of epithelial cell differentiation
mammary gland development
response to nutrient levels
positive regulation of natural killer cell differentiation
negative regulation of mast cell apoptotic process
T cell differentiation in thymus
prolactin signaling pathway
regulation of multicellular organism growth
positive regulation of multicellular organism growth
positive regulation of activated T cell proliferation
T cell homeostasis
negative regulation of apoptotic process
response to peptide hormone
positive regulation of interleukin-2 biosynthetic process
response to ethanol
positive regulation of B cell differentiation
positive regulation of gamma-delta T cell differentiation
positive regulation of lymphocyte differentiation
negative regulation of erythrocyte differentiation
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
positive regulation of natural killer cell mediated cytotoxicity
creatinine metabolic process
development of secondary female sexual characteristics
development of secondary male sexual characteristics
Peyer's patch development
multicellular organismal reproductive process
positive regulation of inflammatory response
mammary gland involution
positive regulation of mast cell differentiation
JAK-STAT cascade involved in growth hormone signaling pathway
prostate gland epithelium morphogenesis
epithelial cell differentiation involved in prostate gland development
mammary gland epithelium development
positive regulation of mast cell proliferation
cellular response to granulocyte macrophage colony-stimulating factor stimulus
RNA polymerase II core promoter sequence-specific DNA binding
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
signal transducer activity
protein binding
protein phosphatase binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR1000014519.rn.1
TC1000001639.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1000014520.rn.1
TC1000001639.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1000014522.rn.1
TC1000001639.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1000014521.rn.1
TC1000001639.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline