Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01497536_g1
          See other KLHL4 GE Assays ›
          Gene Symbol:
          Klhl4
          Gene Name:
          kelch-like family member 4
          Gene Aliases:
          RGD1565441
          Chromosome Location:
          Chr.X: 86125917 - 86229565 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01497536_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klhl4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108244.1 NP_001101714.1 10 - 11 2097 74
          XM_006257199.2 XP_006257261.1 10 - 11 2508 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000800.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P778518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20897 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47751 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49231 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8742 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7979 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100831 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150052 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136535 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P778518 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P778518 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P778518 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P391772 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P375412 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P778518 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73789 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68778 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22939 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10934763 - -
          Affymetrix Rat Gene 1.0 ST Array 10934762 - -
          Affymetrix Rat Gene 1.0 ST Array 10934767 - -
          Affymetrix Rat Gene 1.0 ST Array 10934765 - -
          Affymetrix Rat Gene 1.0 ST Array 10934757 - -
          Affymetrix Rat Gene 1.0 ST Array 10934764 - -
          Affymetrix Rat Gene 1.0 ST Array 10934766 - -
          Affymetrix Rat Gene 1.0 ST Array 10934761 - -
          Affymetrix Rat Gene 1.0 ST Array 10934760 - -
          Affymetrix Rat Gene 1.0 ST Array 10934759 - -
          Affymetrix Rat Gene 1.0 ST Array 10934758 - -
          Affymetrix Rat Gene 1.1 ST Array 10934754 - -
          Rat Genome 230 2.0 Array 1393589_at - -
          Rat Genome 230 2.0 Array 1393589_PM_at - -
          Affymetrix HT Rat Focus Array 1393589_PM_at - -
          Affymetrix HT Rat Focus Array 1393589_at - -
          Affymetrix Rat Clariom S Assay PSR0X00004906.rn.1 TC0X00000800.rn.1 RTA TC
          Rat Expression Set 230 1393589_at - -
          Rat Expression Set 230 1393589_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10934754 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20753906 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650662 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline