Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01497974_m1
          See other DPT GE Assays ›
          Gene Symbol:
          Dpt
          Gene Name:
          dermatopontin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 83073307 - 83102404 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01497974_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dpt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105965.1 NP_001099435.1 1 - 2 400 71
          XM_006250153.1 XP_006250215.1 1 - 2 637 71
          XM_006250154.1 XP_006250216.1 1 - 2 637 71
          XM_008769624.1 XP_008767846.1 1 - 2 637 71
          GenBank mRNA BC167054.1 - 1 - 2 395 7113939 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000660.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s)

          negative regulation of cell proliferation
          collagen fibril organization

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P180259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16266 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168453 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58137 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42290 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P180259 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P180259 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P180259 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P180259 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87294 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10765283 - -
          Affymetrix Rat Gene 1.0 ST Array 10765281 - -
          Affymetrix Rat Gene 1.0 ST Array 10765284 - -
          Affymetrix Rat Gene 1.0 ST Array 10765282 - -
          Affymetrix Rat Gene 1.1 ST Array 10765280 - -
          Rat Genome 230 2.0 Array 1371732_PM_at - -
          Rat Genome 230 2.0 Array 1373947_PM_at - -
          Rat Genome 230 2.0 Array 1371732_at - -
          Rat Genome 230 2.0 Array 1373947_at - -
          Affymetrix HT Rat Focus Array 1371732_PM_at - -
          Affymetrix HT Rat Focus Array 1371732_at - -
          Affymetrix HT Rat Focus Array 1373947_at - -
          Affymetrix HT Rat Focus Array 1373947_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300004631.rn.1 TC1300000660.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004632.rn.1 TC1300000660.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004633.rn.1 TC1300000660.rn.1 RTA TC
          Rat Expression Set 230 1371732_PM_at - -
          Rat Expression Set 230 1373947_at - -
          Rat Expression Set 230 1373947_PM_at - -
          Rat Expression Set 230 1371732_at - -
          Affymetrix Rat Gene 1.0 ST Array 10765280 - -
          Rat Genome U34 Set rc_AI177353_at - -
          Rat Genome U34 Set rc_AI138095_at - -
          Rat Genome U34 Set rc_AI236402_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22396404 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374030 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline