Search
Search
| Gene Symbol: Dpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250153.1 | XP_006250215.1 | 3 - 4 | 870 | 66 | |
| XM_006250154.1 | XP_006250216.1 | 3 - 4 | 870 | 66 | ||
| XM_008769624.1 | XP_008767846.1 | 3 - 4 | 870 | 66 | ||
| GenBank mRNA | BC167054.1 | - | 3 - 4 | 628 | 7113939 | 66 |
Set Membership: |
|