Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01499359_g1
          See other PGAM5 GE Assays ›
          Gene Symbol:
          Pgam5
          Gene Name:
          phosphoglycerate mutase family member 5
          Gene Aliases:
          RGD1312028
          Chromosome Location:
          Chr.12: 52467418 - 52474511 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01499359_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pgam5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025272.1 NP_001020443.1 5 - 6 745 95
          XM_006249534.2 XP_006249596.1 5 - 6 774 95
          GenBank mRNA BC092607.1 - 5 - 6 745 7309810 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pgam5

          Panther Classification:

          Molecular Function -

          isomerase mutase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          dephosphorylation
          positive regulation of GTPase activity
          necroptotic process
          phosphoprotein phosphatase activity
          GTPase activator activity
          phosphatase activity
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P110093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769088 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P110093 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P110093 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P110093 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P370255 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99278 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10759412 - -
          Affymetrix Rat Gene 1.0 ST Array 10759415 - -
          Affymetrix Rat Gene 1.0 ST Array 10759414 - -
          Affymetrix Rat Gene 1.0 ST Array 10759411 - -
          Affymetrix Rat Gene 1.0 ST Array 10759413 - -
          Affymetrix Rat Gene 1.0 ST Array 10759410 - -
          Affymetrix Rat Gene 1.1 ST Array 10759409 - -
          Rat Genome 230 2.0 Array 1391577_PM_at - -
          Rat Genome 230 2.0 Array 1391577_at - -
          Rat Genome 230 2.0 Array 1378382_at - -
          Rat Genome 230 2.0 Array 1378382_PM_at - -
          Affymetrix HT Rat Focus Array 1391577_at - -
          Affymetrix HT Rat Focus Array 1378382_PM_at - -
          Affymetrix HT Rat Focus Array 1391577_PM_at - -
          Affymetrix HT Rat Focus Array 1378382_at - -
          Affymetrix Rat Clariom S Assay PSR1200007816.rn.1 TC1200000881.rn.1 RTA TC
          Rat Expression Set 230 1378382_at - -
          Rat Expression Set 230 1378382_PM_at - -
          Rat Expression Set 230 1391577_at - -
          Rat Expression Set 230 1391577_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10759409 - -
          Rat Genome U34 Set rc_AI230014_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20857117 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369510 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline