Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01500035_g1
          See other LOC304396 GE Assays ›
          Gene Symbol:
          LOC304396
          Gene Name:
          similar to hypothetical protein DKFZp434K1815
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 23574233 - 23587262 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01500035_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC304396
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013981.1 NP_001014003.1 11 - 12 2025 95
          XM_006249156.2 XP_006249218.1 12 - 13 1636 95
          GenBank mRNA BC081940.1 - 11 - 12 2025 7115744 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000401.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC304396

          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          transcription, DNA-templated
          protein phosphorylation
          cell proliferation
          cytokine-mediated signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          establishment of protein localization to chromatin
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          signal transducer activity
          methyl-CpG binding
          methylated histone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591364 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159986 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038772 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038772 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038772 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P220008 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33904 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757574 - -
          Affymetrix Rat Gene 1.0 ST Array 10757579 - -
          Affymetrix Rat Gene 1.0 ST Array 10757584 - -
          Affymetrix Rat Gene 1.0 ST Array 10757580 - -
          Affymetrix Rat Gene 1.0 ST Array 10757581 - -
          Affymetrix Rat Gene 1.0 ST Array 10757569 - -
          Affymetrix Rat Gene 1.0 ST Array 10757576 - -
          Affymetrix Rat Gene 1.0 ST Array 10757577 - -
          Affymetrix Rat Gene 1.0 ST Array 10757583 - -
          Affymetrix Rat Gene 1.0 ST Array 10757578 - -
          Affymetrix Rat Gene 1.0 ST Array 10757585 - -
          Affymetrix Rat Gene 1.0 ST Array 10757575 - -
          Affymetrix Rat Gene 1.0 ST Array 10757582 - -
          Affymetrix Rat Gene 1.0 ST Array 10757568 - -
          Affymetrix Rat Gene 1.0 ST Array 10757573 - -
          Affymetrix Rat Gene 1.0 ST Array 10757570 - -
          Affymetrix Rat Gene 1.0 ST Array 10757567 - -
          Affymetrix Rat Gene 1.0 ST Array 10757572 - -
          Affymetrix Rat Gene 1.1 ST Array 10757566 - -
          Rat Genome 230 2.0 Array 1373715_PM_at - -
          Rat Genome 230 2.0 Array 1373715_at - -
          Affymetrix HT Rat Focus Array 1373715_PM_at - -
          Affymetrix HT Rat Focus Array 1373715_at - -
          Affymetrix Rat Clariom S Assay PSR1200003589.rn.1 TC1200000401.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003586.rn.1 TC1200000401.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003588.rn.1 TC1200000401.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003587.rn.1 TC1200000401.rn.1 RTA TC
          Rat Expression Set 230 1373715_at - -
          Rat Expression Set 230 1373715_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757566 - -
          Rat Genome U34 Set rc_AA893517_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21676615 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376647 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375440 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline