Search
Search
| Gene Symbol: Tac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124769.1 | NP_001118241.1 | 4 - 5 | 336 | 93 | |
| NM_001124770.1 | NP_001118242.1 | 5 - 6 | 381 | 138 | ||
| XM_008762694.1 | XP_008760916.1 | 5 - 6 | 525 | 138 | ||
| XM_008762695.1 | XP_008760917.1 | 4 - 5 | 478 | 93 | ||
| GenBank mRNA | M34184.1 | - | 5 - 6 | 383 | 138 | |
| X56306.1 | - | 3 - 4 | 243 | 93 | ||
Set Membership: |
|