Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01500473_m1
          See other PTPRZ1 GE Assays ›
          Gene Symbol:
          Ptprz1
          Gene Name:
          protein tyrosine phosphatase, receptor-type, Z polypeptide 1
          Gene Aliases:
          PTPzeta-A, PTPzeta-B, PTPzeta-S, Ptpz, RPTPbeta
          Chromosome Location:
          Chr.4: 49941265 - 50140764 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01500473_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptprz1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170685.1 NP_001164156.1 11 - 12 1392 87
          NM_013080.2 NP_037212.2 11 - 12 1392 87
          XM_006236137.2 XP_006236199.1 11 - 12 1431 87
          XM_006236138.2 XP_006236200.1 11 - 12 1431 87
          XM_006236139.2 XP_006236201.1 11 - 12 1431 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000467.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase 3' Most Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptprz1

          Panther Classification:

          Molecular Function -

          protein phosphatase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          protein dephosphorylation
          axonogenesis
          axonal fasciculation
          brain development
          learning or memory
          negative regulation of cell proliferation
          visual learning
          negative regulation of cell-substrate adhesion
          positive regulation of neuron projection development
          hippocampus development
          positive regulation of cell migration
          peptidyl-tyrosine dephosphorylation
          positive regulation of fibroblast proliferation
          neuron development
          oligodendrocyte differentiation
          regulation of dendrite morphogenesis
          positive regulation of peptidyl-tyrosine phosphorylation
          regulation of oligodendrocyte progenitor proliferation
          positive regulation of dendrite development
          positive regulation of Schwann cell migration
          positive regulation of neuron death
          negative regulation of dendrite development
          positive regulation of neuron migration
          protein tyrosine phosphatase activity
          protein binding
          phosphatase activity
          fibroblast growth factor binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P129263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276656 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155933 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119122 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25661 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49860 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42182 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26258 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95905 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72893 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17481 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94745 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123115 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140885 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64753 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163317 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75530 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15436 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103001 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41143 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123863 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33304 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4952 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129263 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129263 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P129263 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P482128 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P475885 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700497354 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88400 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99207 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92265 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44040 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29832 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77688 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5022 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15474 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24242 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17307 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62548 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96421 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38625 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52552 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99232 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88009 - -
          Agilent Rat cDNA Microarray (G4105A) 700497354 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10853964 - -
          Affymetrix Rat Gene 1.0 ST Array 10853983 - -
          Affymetrix Rat Gene 1.0 ST Array 10853990 - -
          Affymetrix Rat Gene 1.0 ST Array 10853968 - -
          Affymetrix Rat Gene 1.0 ST Array 10853994 - -
          Affymetrix Rat Gene 1.0 ST Array 10853967 - -
          Affymetrix Rat Gene 1.0 ST Array 10853974 - -
          Affymetrix Rat Gene 1.0 ST Array 10853986 - -
          Affymetrix Rat Gene 1.0 ST Array 10853980 - -
          Affymetrix Rat Gene 1.0 ST Array 10853965 - -
          Affymetrix Rat Gene 1.0 ST Array 10853984 - -
          Affymetrix Rat Gene 1.0 ST Array 10853979 - -
          Affymetrix Rat Gene 1.0 ST Array 10853988 - -
          Affymetrix Rat Gene 1.0 ST Array 10853972 - -
          Affymetrix Rat Gene 1.0 ST Array 10853989 - -
          Affymetrix Rat Gene 1.0 ST Array 10853970 - -
          Affymetrix Rat Gene 1.0 ST Array 10853977 - -
          Affymetrix Rat Gene 1.0 ST Array 10853982 - -
          Affymetrix Rat Gene 1.0 ST Array 10853971 - -
          Affymetrix Rat Gene 1.0 ST Array 10853975 - -
          Affymetrix Rat Gene 1.0 ST Array 10853978 - -
          Affymetrix Rat Gene 1.0 ST Array 10853973 - -
          Affymetrix Rat Gene 1.0 ST Array 10853966 - -
          Affymetrix Rat Gene 1.0 ST Array 10853981 - -
          Affymetrix Rat Gene 1.0 ST Array 10853969 - -
          Affymetrix Rat Gene 1.0 ST Array 10853987 - -
          Affymetrix Rat Gene 1.0 ST Array 10853992 - -
          Affymetrix Rat Gene 1.0 ST Array 10853993 - -
          Affymetrix Rat Gene 1.0 ST Array 10853985 - -
          Affymetrix Rat Gene 1.0 ST Array 10853976 - -
          Affymetrix Rat Gene 1.0 ST Array 10853991 - -
          Affymetrix Rat Gene 1.1 ST Array 10853963 - -
          Rat Genome 230 2.0 Array 1368350_at - -
          Rat Genome 230 2.0 Array 1368350_PM_at - -
          Rat Genome 230 2.0 Array 1388015_at - -
          Rat Genome 230 2.0 Array 1388015_PM_at - -
          Affymetrix HT Rat Focus Array 1368350_at - -
          Affymetrix HT Rat Focus Array 1368350_PM_at - -
          Affymetrix HT Rat Focus Array 1388015_PM_at - -
          Affymetrix HT Rat Focus Array 1388015_at - -
          Affymetrix Rat Clariom S Assay PSR0400003313.rn.1 TC0400000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003314.rn.1 TC0400000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003315.rn.1 TC0400000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003312.rn.1 TC0400000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003316.rn.1 TC0400000467.rn.1 RTA TC
          Rat Expression Set 230 1388015_PM_at - -
          Rat Expression Set 230 1388015_at - -
          Rat Expression Set 230 1368350_PM_at - -
          Rat Expression Set 230 1368350_at - -
          Affymetrix Rat Gene 1.0 ST Array 10853963 - -
          Rat Genome U34 Set U04998_at - -
          Rat Genome U34 Set U09357_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline