Search
Search
| Gene Symbol: Irf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106586.1 | NP_001100056.1 | 6 - 7 | 825 | 71 | |
| XM_006236180.2 | XP_006236242.1 | 5 - 6 | 1978 | 71 | ||
| XM_006236183.2 | XP_006236245.1 | 6 - 7 | 1121 | 71 | ||
| XM_006236184.2 | XP_006236246.1 | 6 - 7 | 861 | 71 | ||
| XM_006236185.2 | XP_006236247.1 | 6 - 7 | 1912 | 71 | ||
| XM_008762765.1 | XP_008760987.1 | 6 - 7 | 778 | 71 | ||
| GenBank mRNA | FQ227572.1 | - | 6 - 7 | 817 | 71 | |
| FQ230949.1 | - | 1 - 2 | 46 | 71 | ||
Set Membership: |
|