Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01500560_m1
          See other BRAF GE Assays ›
          Gene Symbol:
          Braf
          Gene Name:
          B-Raf proto-oncogene, serine/threonine kinase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 67389331 - 67520487 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01500560_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Braf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006236357.2 XP_006236419.2 19 - 20 2389 135
          XM_006236358.2 XP_006236420.2 19 - 20 2305 135
          XM_006236359.2 XP_006236421.2 18 - 19 2269 135
          XM_231692.8 XP_231692.4 21 - 22 2431 135

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Apoptosis Protein Kinase 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          myeloid progenitor cell differentiation
          protein phosphorylation
          visual learning
          positive regulation of gene expression
          negative regulation of fibroblast migration
          cell differentiation
          positive regulation of peptidyl-serine phosphorylation
          somatic stem cell population maintenance
          cellular response to drug
          regulation of cell proliferation
          negative regulation of apoptotic process
          CD4-positive, alpha-beta T cell differentiation
          positive T cell selection
          response to peptide hormone
          negative regulation of neuron apoptotic process
          alpha-beta T cell differentiation
          regulation of axon regeneration
          positive regulation of axon regeneration
          positive regulation of axonogenesis
          protein heterooligomerization
          positive regulation of stress fiber assembly
          response to cAMP
          long-term synaptic potentiation
          head morphogenesis
          positive regulation of ERK1 and ERK2 cascade
          cellular response to calcium ion
          positive regulation of substrate adhesion-dependent cell spreading
          negative regulation of synaptic vesicle exocytosis
          negative regulation of endothelial cell apoptotic process
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          calcium ion binding
          protein binding
          ATP binding
          small GTPase binding
          mitogen-activated protein kinase kinase binding
          identical protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P16155 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16155 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16155 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P16155 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16155 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2704 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10862088 - -
          Affymetrix Rat Gene 1.0 ST Array 10862085 - -
          Affymetrix Rat Gene 1.0 ST Array 10862089 - -
          Affymetrix Rat Gene 1.0 ST Array 10862086 - -
          Affymetrix Rat Gene 1.0 ST Array 10862087 - -
          Affymetrix Rat Gene 1.1 ST Array 10862082 - -
          Affymetrix Rat Gene 1.0 ST Array 10862082 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21315367 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358239 - -
          RatRef-12 Expression BeadChip ILMN_53294 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline