Search
Search
| Gene Symbol: Mdfic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105668.1 | NP_001099138.1 | 2 - 3 | 454 | 146 | |
| XM_006236103.2 | XP_006236165.1 | 2 - 3 | 759 | 146 | ||
| GenBank mRNA | CV106654.1 | - | 255 | 146 | ||
Set Membership: |
|