Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01501045_g1
          See other CAPZA2 GE Assays ›
          Gene Symbol:
          Capza2
          Gene Name:
          capping protein (actin filament) muscle Z-line, alpha 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 44936290 - 44972270 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01501045_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Capza2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009180.2 NP_001009180.1 9 - 10 780 189
          GenBank mRNA BC101867.1 - 9 - 10 780 7938377 189

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000433.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Capza2

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          barbed-end actin filament capping
          actin binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P354266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511464 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33281 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79240 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P354266 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P354266 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P354266 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P354266 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10924 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77252 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42026 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67945 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75318 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10853847 - -
          Affymetrix Rat Gene 1.0 ST Array 10853849 - -
          Affymetrix Rat Gene 1.0 ST Array 10853848 - -
          Affymetrix Rat Gene 1.0 ST Array 10853845 - -
          Affymetrix Rat Gene 1.0 ST Array 10853843 - -
          Affymetrix Rat Gene 1.0 ST Array 10853850 - -
          Affymetrix Rat Gene 1.0 ST Array 10853846 - -
          Affymetrix Rat Gene 1.0 ST Array 10853851 - -
          Affymetrix Rat Gene 1.0 ST Array 10853844 - -
          Affymetrix Rat Gene 1.1 ST Array 10853842 - -
          Rat Genome 230 2.0 Array 1399043_at - -
          Rat Genome 230 2.0 Array 1399043_PM_at - -
          Affymetrix HT Rat Focus Array 1399043_PM_at - -
          Affymetrix HT Rat Focus Array 1399043_at - -
          Affymetrix Rat Clariom S Assay PSR0400003108.rn.1 TC0400000433.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003104.rn.1 TC0400000433.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003107.rn.1 TC0400000433.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003105.rn.1 TC0400000433.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003106.rn.1 TC0400000433.rn.1 RTA TC
          Rat Expression Set 230 1399043_at - -
          Rat Expression Set 230 1399043_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10853842 - -
          Rat Genome U34 Set rc_AA892842_at - -
          Rat Genome U34 Set rc_AI175266_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372330 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline