Search
Search
| Gene Symbol: Drap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077668.2 | NP_001071136.1 | 1 - 2 | 251 | 75 | |
| XM_006230741.1 | XP_006230803.1 | 1 - 2 | 294 | 75 | ||
| GenBank mRNA | BC127524.1 | - | 1 - 2 | 252 | 7114598 | 75 |
| FQ231535.1 | - | 1 - 2 | 117 | 75 | ||
Set Membership: |
|