Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01502065_g1
          See other FERMT3 GE Assays ›
          Gene Symbol:
          Fermt3
          Gene Name:
          fermitin family member 3
          Gene Aliases:
          RGD1310168
          Chromosome Location:
          Chr.1: 222254178 - 222272779 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01502065_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fermt3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127543.1 NP_001121015.1 4 - 5 626 98
          XM_006230811.2 XP_006230873.1 4 - 5 1117 98
          GenBank mRNA BC166607.1 - 4 - 5 626 5598287 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007186.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          leukocyte cell-cell adhesion
          integrin-mediated signaling pathway
          integrin activation
          regulation of cell-cell adhesion mediated by integrin
          substrate adhesion-dependent cell spreading
          platelet aggregation
          integrin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P276439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910257 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42147 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098454 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098454 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P098454 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P492104 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93362 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72642 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728356 - -
          Affymetrix Rat Gene 1.0 ST Array 10728348 - -
          Affymetrix Rat Gene 1.0 ST Array 10728358 - -
          Affymetrix Rat Gene 1.0 ST Array 10728352 - -
          Affymetrix Rat Gene 1.0 ST Array 10728347 - -
          Affymetrix Rat Gene 1.0 ST Array 10728355 - -
          Affymetrix Rat Gene 1.0 ST Array 10728357 - -
          Affymetrix Rat Gene 1.0 ST Array 10728354 - -
          Affymetrix Rat Gene 1.0 ST Array 10728351 - -
          Affymetrix Rat Gene 1.0 ST Array 10728359 - -
          Affymetrix Rat Gene 1.0 ST Array 10728353 - -
          Affymetrix Rat Gene 1.0 ST Array 10728350 - -
          Affymetrix Rat Gene 1.0 ST Array 10728349 - -
          Affymetrix Rat Gene 1.0 ST Array 10728360 - -
          Affymetrix Rat Gene 1.1 ST Array 10728346 - -
          Rat Genome 230 2.0 Array 1388740_at - -
          Rat Genome 230 2.0 Array 1384879_at - -
          Rat Genome 230 2.0 Array 1388740_PM_at - -
          Rat Genome 230 2.0 Array 1384879_PM_at - -
          Affymetrix HT Rat Focus Array 1388740_at - -
          Affymetrix HT Rat Focus Array 1388740_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100057852.rn.1 TC0100007186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057863.rn.1 TC0100007186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057859.rn.1 TC0100007186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057850.rn.1 TC0100007186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057853.rn.1 TC0100007186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057861.rn.1 TC0100007186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057854.rn.1 TC0100007186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057855.rn.1 TC0100007186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057856.rn.1 TC0100007186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057857.rn.1 TC0100007186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057864.rn.1 TC0100007186.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100057865.rn.1 TC0100007186.rn.1 RTA TC
          Rat Expression Set 230 1388740_PM_at - -
          Rat Expression Set 230 1388740_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728346 - -
          Rat Genome U34 Set rc_AI177645_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21027949 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351911 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline