Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01502299_g1
          See other EIF3L GE Assays ›
          Gene Symbol:
          Eif3l
          Gene Name:
          eukaryotic translation initiation factor 3, subunit L
          Gene Aliases:
          Eif3s6ip
          Chromosome Location:
          Chr.7: 120320547 - 120331596 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01502299_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eif3l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034134.1 NP_001029306.1 5 - 6 1230 60
          GenBank mRNA BC100264.1 - 5 - 6 1230 7382342 60
          FQ209474.1 - 1254 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001442.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eif3l

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          formation of translation preinitiation complex
          translational initiation
          regulation of translational initiation
          translation initiation factor activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P222878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77238 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46496 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P222878 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P222878 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P222878 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P699937 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P222878 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65804 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897601 - -
          Affymetrix Rat Gene 1.0 ST Array 10897599 - -
          Affymetrix Rat Gene 1.0 ST Array 10897598 - -
          Affymetrix Rat Gene 1.0 ST Array 10897603 - -
          Affymetrix Rat Gene 1.0 ST Array 10897602 - -
          Affymetrix Rat Gene 1.0 ST Array 10897600 - -
          Affymetrix Rat Gene 1.1 ST Array 10897590 - -
          Rat Genome 230 2.0 Array 1398929_PM_at - -
          Rat Genome 230 2.0 Array 1390880_PM_at - -
          Rat Genome 230 2.0 Array 1398929_at - -
          Rat Genome 230 2.0 Array 1390880_at - -
          Affymetrix HT Rat Focus Array 1390880_PM_at - -
          Affymetrix HT Rat Focus Array 1398929_PM_at - -
          Affymetrix HT Rat Focus Array 1390880_at - -
          Affymetrix HT Rat Focus Array 1398929_at - -
          Affymetrix Rat Clariom S Assay PSR0700010749.rn.1 TC0700001442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010748.rn.1 TC0700001442.rn.1 RTA TC
          Rat Expression Set 230 1398929_at - -
          Rat Expression Set 230 1398929_PM_at - -
          Rat Expression Set 230 1390880_PM_at - -
          Rat Expression Set 230 1390880_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897590 - -
          Rat Genome U34 Set rc_AA955240_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21217114 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline