Search
Search
| Gene Symbol: Ccdc38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241271.2 | XP_006241333.1 | 17 - 18 | 1935 | 123 | |
| XM_008765310.1 | XP_008763532.1 | 16 - 17 | 1665 | 123 | ||
Set Membership: |
|