Search
Search
| Gene Symbol: Appl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108741.1 | NP_001102211.1 | 7 - 8 | 555 | 66 | |
| XM_006241201.2 | XP_006241263.1 | 7 - 8 | 596 | 66 | ||
| GenBank mRNA | BC168167.1 | - | 7 - 8 | 470 | 9021772 | 66 |
Set Membership: |
|