Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01503688_m1
          See other IGF1 GE Assays ›
          Gene Symbol:
          Igf1
          Gene Name:
          insulin-like growth factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 28412123 - 28491815 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01503688_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001082477.2 NP_001075946.2 4 - 5 429 75
          NM_001082478.1 NP_001075947.1 4 - 5 726 75
          XM_006241191.2 XP_006241253.1 5 - 6 570 75
          XM_006241192.1 XP_006241254.1 5 - 6 484 75
          XM_006241194.2 XP_006241256.1 4 - 5 608 75
          GenBank mRNA FQ210560.1 - 4 - 5 442 75
          FQ220385.1 - 4 - 5 750 75
          M15480.1 - 4 - 5 473 75
          X06107.1 - 4 - 5 704 75
          X06108.1 - 5 - 6 518 75

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell activation
          regulation of protein phosphorylation
          blood vessel remodeling
          regulation of translation
          nervous system development
          female pregnancy
          memory
          cell proliferation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          response to heat
          response to mechanical stimulus
          glial cell differentiation
          response to organic substance
          regulation of gene expression
          positive regulation of cardiac muscle hypertrophy
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to organic cyclic compound
          response to activity
          skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
          muscle hypertrophy
          myotube cell development
          positive regulation of smooth muscle cell migration
          positive regulation of cerebellar granule cell precursor proliferation
          water homeostasis
          proteoglycan biosynthetic process
          positive regulation of cell growth
          lung development
          mammary gland development
          exocrine pancreas development
          response to nutrient levels
          response to lipopolysaccharide
          regulation of establishment or maintenance of cell polarity
          positive regulation of protein import into nucleus, translocation
          negative regulation of smooth muscle cell apoptotic process
          multicellular organism growth
          bone mineralization involved in bone maturation
          regulation of multicellular organism growth
          positive regulation of activated T cell proliferation
          regulation of cell proliferation
          positive regulation of tyrosine phosphorylation of Stat5 protein
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of DNA binding
          positive regulation of MAPK cascade
          positive regulation of insulin-like growth factor receptor signaling pathway
          myoblast differentiation
          positive regulation of fat cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of glycogen biosynthetic process
          positive regulation of DNA replication
          positive regulation of glycolytic process
          positive regulation of mitotic nuclear division
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of glucose import
          positive regulation of Ras protein signal transduction
          insulin-like growth factor receptor signaling pathway
          phosphatidylinositol-mediated signaling
          positive regulation of fibroblast proliferation
          lung alveolus development
          cell development
          response to steroid hormone
          positive regulation of smooth muscle cell proliferation
          branching morphogenesis of an epithelial tube
          inner ear development
          chondroitin sulfate proteoglycan biosynthetic process
          positive regulation of epithelial cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          detection of mechanical stimulus involved in sensory perception
          regulation of protein metabolic process
          response to glucocorticoid
          myoblast proliferation
          positive regulation of protein kinase B signaling
          regulation of calcium ion transport
          negative regulation of oocyte development
          lung vasculature development
          lung lobe morphogenesis
          Type I pneumocyte differentiation
          Type II pneumocyte differentiation
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          prostate gland growth
          prostate gland epithelium morphogenesis
          prostate gland stromal morphogenesis
          negative regulation of androgen receptor signaling pathway
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of calcineurin-NFAT signaling cascade
          cellular response to electrical stimulus
          positive regulation of protein serine/threonine kinase activity
          positive regulation of steroid hormone biosynthetic process
          negative regulation of release of cytochrome c from mitochondria
          extrinsic apoptotic signaling pathway in absence of ligand
          negative regulation of neuron death
          positive regulation of myoblast proliferation
          negative regulation of extrinsic apoptotic signaling pathway
          insulin receptor binding
          insulin-like growth factor receptor binding
          integrin binding
          hormone activity
          steroid binding
          protein binding
          growth factor activity
          protein serine/threonine kinase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052742 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P366723 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324733 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128654 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74784 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175417 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43513 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80736 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15842 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24865 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P366723 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052740 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P366723 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052740 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052740 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P366723 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P421499 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P421502 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P366723 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P126021 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700695253 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98820 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38869 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40809 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87199 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49433 - -
          Agilent Rat cDNA Microarray (G4105A) 700695253 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10894701 - -
          Affymetrix Rat Gene 1.0 ST Array 10894696 - -
          Affymetrix Rat Gene 1.0 ST Array 10894703 - -
          Affymetrix Rat Gene 1.0 ST Array 10894697 - -
          Affymetrix Rat Gene 1.0 ST Array 10894698 - -
          Affymetrix Rat Gene 1.0 ST Array 10894704 - -
          Affymetrix Rat Gene 1.0 ST Array 10894700 - -
          Affymetrix Rat Gene 1.0 ST Array 10894699 - -
          Affymetrix Rat Gene 1.1 ST Array 10894695 - -
          Rat Genome 230 2.0 Array 1370333_PM_a_at - -
          Rat Genome 230 2.0 Array 1370333_a_at - -
          Affymetrix HT Rat Focus Array 1370333_a_at - -
          Affymetrix HT Rat Focus Array 1370333_PM_a_at - -
          Rat Expression Set 230 1370333_a_at - -
          Rat Expression Set 230 1370333_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10894695 - -
          Rat Genome U34 Set M15481_at - -
          Rat Genome U34 Set D00698_s_at - -
          Rat Genome U34 Set M15481_g_at - -
          Rat Genome U34 Set X06107_i_at - -
          Rat Genome U34 Set X06107_r_at - -
          Rat Genome U34 Set X06107_i_at - -
          Rat Genome U34 Set D00698_s_at - -
          Rat Genome U34 Set M15481_g_at - -
          Rat Genome U34 Set X06107_r_at - -
          Rat Genome U34 Set M15481_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21999055 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline