Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01504238_m1
          See other GLI1 GE Assays ›
          Gene Symbol:
          Gli1
          Gene Name:
          GLI family zinc finger 1
          Gene Aliases:
          Gli
          Chromosome Location:
          Chr.7: 70620794 - 70633163 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01504238_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gli1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191910.1 NP_001178839.1 10 - 11 1573 55
          XM_006241442.2 XP_006241504.1 12 - 13 1838 55
          XM_006241443.2 XP_006241505.1 11 - 12 1721 55
          XM_008765375.1 XP_008763597.1 12 - 13 1786 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002918.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          transcription from RNA polymerase II promoter
          smoothened signaling pathway
          spermatogenesis
          ventral midline development
          biological_process
          positive regulation of cell proliferation
          epidermal cell differentiation
          dorsal/ventral pattern formation
          proximal/distal pattern formation
          cerebellar cortex morphogenesis
          smoothened signaling pathway involved in regulation of cerebellar granule cell precursor cell proliferation
          pituitary gland development
          lung development
          positive regulation of DNA replication
          positive regulation of smoothened signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          notochord regression
          negative regulation of canonical Wnt signaling pathway
          molecular_function
          DNA binding
          chromatin binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          microtubule binding
          transcription regulatory region DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1054991 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1054991 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1054991 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1054991 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16047 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1054993 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1054991 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903038 - -
          Affymetrix Rat Gene 1.1 ST Array 10903035 - -
          Rat Genome 230 2.0 Array 1378720_at - -
          Rat Genome 230 2.0 Array 1378720_PM_at - -
          Affymetrix HT Rat Focus Array 1378720_PM_at - -
          Affymetrix HT Rat Focus Array 1378720_at - -
          Affymetrix Rat Clariom S Assay PSR0700022574.rn.1 TC0700002918.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022575.rn.1 TC0700002918.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022576.rn.1 TC0700002918.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022573.rn.1 TC0700002918.rn.1 RTA TC
          Rat Expression Set 230 1378720_PM_at - -
          Rat Expression Set 230 1378720_at - -
          Affymetrix Rat Gene 1.0 ST Array 10903035 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21306512 - -
          Array Name Probe IDs Targets Target Type
          RatRef-12 Expression BeadChip ILMN_60990 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline