Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01504551_m1
          See other GTPBP1 GE Assays ›
          Gene Symbol:
          Gtpbp1
          Gene Name:
          GTP binding protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 120923274 - 120947723 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01504551_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gtpbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199315.1 NP_001186244.1 11 - 12 1964 98
          GenBank mRNA GU256773.1 - 11 - 12 1924 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001457.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          translation elongation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          positive regulation of mRNA catabolic process
          GTPase activity
          GTP binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113475 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38506 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28742 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038329 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038329 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038329 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20103 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897719 - -
          Affymetrix Rat Gene 1.0 ST Array 10897718 - -
          Affymetrix Rat Gene 1.0 ST Array 10897720 - -
          Affymetrix Rat Gene 1.0 ST Array 10897711 - -
          Affymetrix Rat Gene 1.0 ST Array 10897710 - -
          Affymetrix Rat Gene 1.1 ST Array 10897709 - -
          Rat Genome 230 2.0 Array 1398985_PM_at - -
          Rat Genome 230 2.0 Array 1375447_PM_at - -
          Rat Genome 230 2.0 Array 1398985_at - -
          Rat Genome 230 2.0 Array 1379905_PM_at - -
          Rat Genome 230 2.0 Array 1379905_at - -
          Rat Genome 230 2.0 Array 1375447_at - -
          Affymetrix HT Rat Focus Array 1398985_at - -
          Affymetrix HT Rat Focus Array 1375447_at - -
          Affymetrix HT Rat Focus Array 1375447_PM_at - -
          Affymetrix HT Rat Focus Array 1398985_PM_at - -
          Affymetrix HT Rat Focus Array 1379905_PM_at - -
          Affymetrix HT Rat Focus Array 1379905_at - -
          Affymetrix Rat Clariom S Assay PSR0700010957.rn.1 TC0700001457.rn.1 RTA TC
          Rat Expression Set 230 1375447_PM_at - -
          Rat Expression Set 230 1398985_PM_at - -
          Rat Expression Set 230 1398985_at - -
          Rat Expression Set 230 1375447_at - -
          Rat Expression Set 230 1379905_PM_at - -
          Rat Expression Set 230 1379905_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897709 - -
          Rat Genome U34 Set rc_AI072081_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21602042 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349875 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline