Search
Search

Laptm4b| Gene Symbol: Laptm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013174.1 | NP_001013192.1 | 2 - 3 | 282 | 127 | |
| GenBank mRNA | BC086442.1 | - | 2 - 3 | 282 | 7303600 | 127 |
Set Membership: |
|