Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505756_m1
          See other NRBP2 GE Assays ›
          Gene Symbol:
          Nrbp2
          Gene Name:
          nuclear receptor binding protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 117145106 - 117151694 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505756_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nrbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135007.1 NP_001128479.1 13 - 14 1704 79
          XM_006241885.2 XP_006241947.1 14 - 15 1723 79
          GenBank mRNA FQ223130.1 - 8 - 9 1265 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003382.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          negative regulation of macroautophagy
          neuron differentiation
          negative regulation of neuron apoptotic process
          molecular_function
          protein kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P096826 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P250983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105756 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P250983 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P096826 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P096826 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P250983 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P096826 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P250983 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P191287 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P250983 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10904782 - -
          Affymetrix Rat Gene 1.0 ST Array 10904778 - -
          Affymetrix Rat Gene 1.0 ST Array 10904775 - -
          Affymetrix Rat Gene 1.0 ST Array 10904770 - -
          Affymetrix Rat Gene 1.0 ST Array 10904785 - -
          Affymetrix Rat Gene 1.0 ST Array 10904773 - -
          Affymetrix Rat Gene 1.0 ST Array 10904776 - -
          Affymetrix Rat Gene 1.0 ST Array 10904784 - -
          Affymetrix Rat Gene 1.0 ST Array 10904777 - -
          Affymetrix Rat Gene 1.0 ST Array 10904780 - -
          Affymetrix Rat Gene 1.0 ST Array 10904783 - -
          Affymetrix Rat Gene 1.0 ST Array 10904771 - -
          Affymetrix Rat Gene 1.0 ST Array 10904779 - -
          Affymetrix Rat Gene 1.0 ST Array 10904786 - -
          Affymetrix Rat Gene 1.0 ST Array 10904774 - -
          Affymetrix Rat Gene 1.0 ST Array 10904772 - -
          Affymetrix Rat Gene 1.0 ST Array 10904781 - -
          Affymetrix Rat Gene 1.1 ST Array 10904769 - -
          Rat Genome 230 2.0 Array 1374417_PM_at - -
          Rat Genome 230 2.0 Array 1374417_at - -
          Affymetrix Rat Clariom S Assay PSR0700026071.rn.1 TC0700003382.rn.1 RTA TC
          Rat Expression Set 230 1374417_PM_at - -
          Rat Expression Set 230 1374417_at - -
          Affymetrix Rat Gene 1.0 ST Array 10904769 - -
          Rat Genome U34 Set rc_AI227690_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20709002 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline