Search
Search

Pacsin2| Gene Symbol: Pacsin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006242054.2 | XP_006242116.1 | 7 - 8 | 1461 | 108 | |
| XM_006242057.2 | XP_006242119.1 | 9 - 10 | 1943 | 108 | ||
| XM_006242058.1 | XP_006242120.1 | 9 - 10 | 1191 | 108 | ||
| XM_006242059.2 | XP_006242121.1 | 8 - 9 | 1936 | 108 | ||
| XM_006242060.1 | XP_006242122.1 | 8 - 9 | 1185 | 108 | ||
| GenBank mRNA | AF139493.1 | - | 9 - 10 | 1167 | 108 | |
| AF139495.1 | - | 8 - 9 | 1161 | 108 | ||
Set Membership: |
|