Search
Search
| Gene Symbol: Emc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113785.1 | NP_001107257.1 | 4 - 5 | 387 | 116 | |
| GenBank mRNA | BC158805.1 | - | 4 - 5 | 387 | 7322060 | 116 |
| FQ214096.1 | - | 1 - 2 | 270 | 116 | ||
| FQ215024.1 | - | 4 - 5 | 376 | 116 | ||
Set Membership: |
|