Search
Search
| Gene Symbol: Acsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108502.2 | NP_001101972.2 | 5 - 6 | 869 | 82 | |
| XM_006230127.2 | XP_006230189.1 | 6 - 7 | 1022 | 82 | ||
| XM_006230128.2 | XP_006230190.1 | 6 - 7 | 1019 | 82 | ||
| XM_006230129.2 | XP_006230191.1 | 5 - 6 | 785 | 82 | ||
| XM_008759771.1 | XP_008757993.1 | 6 - 7 | 938 | 82 | ||
| XM_008759772.1 | XP_008757994.1 | 5 - 6 | 914 | 82 | ||
| XM_008759773.1 | XP_008757995.1 | 6 - 7 | 1019 | 82 | ||
| GenBank mRNA | BC168985.1 | - | 5 - 6 | 869 | 7323876 | 82 |
| FQ218416.1 | - | 1 - 2 | 62 | 82 | ||
Set Membership: |
|