Search
Search
| Gene Symbol: Scnn1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017046.1 | NP_058742.1 | 1 - 2 | 63 | 83 | |
| GenBank mRNA | X77933.1 | - | 1 - 2 | 63 | 83 | |
| X78034.1 | - | 1 - 2 | 46 | 83 | ||
Set Membership: |
|