Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01507866_m1
          See other AQP11 GE Assays ›
          Gene Symbol:
          Aqp11
          Gene Name:
          aquaporin 11
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 162703394 - 162713560 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01507866_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aqp11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173105.1 NP_775128.1 2 - 3 964 116
          GenBank mRNA AB023644.1 - 2 - 3 964 116
          BC078694.1 - 2 - 3 991 7111950 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006082.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aqp11

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          ion transport
          water transport
          glycerol transport
          urea transport
          endosomal lumen acidification
          protein homooligomerization
          transmembrane transport
          proximal tubule development
          molecular_function
          transporter activity
          water channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P274083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427977 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56325 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P274083 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P274083 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P274083 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P274083 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13315 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10723730 - -
          Affymetrix Rat Gene 1.0 ST Array 10723732 - -
          Affymetrix Rat Gene 1.0 ST Array 10723733 - -
          Affymetrix Rat Gene 1.0 ST Array 10723731 - -
          Affymetrix Rat Gene 1.0 ST Array 10723729 - -
          Affymetrix Rat Gene 1.1 ST Array 10723728 - -
          Rat Genome 230 2.0 Array 1384877_at - -
          Rat Genome 230 2.0 Array 1384877_PM_at - -
          Affymetrix HT Rat Focus Array 1384877_at - -
          Affymetrix HT Rat Focus Array 1384877_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100048246.rn.1 TC0100006082.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100048245.rn.1 TC0100006082.rn.1 RTA TC
          Rat Expression Set 230 1384877_at - -
          Rat Expression Set 230 1384877_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10723728 - -
          Rat Genome U34 Set rc_AI072405_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21797494 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373630 - -
          RatRef-12 Expression BeadChip ILMN_67993 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline