Search
Search
| Gene Symbol: Lrrc51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106284.1 | NP_001099754.1 | 2 - 3 | 192 | 87 | |
| XM_006229874.2 | XP_006229936.1 | 2 - 3 | 148 | 87 | ||
| GenBank mRNA | EU627092.1 | - | 1 - 2 | 61 | 87 | |
Set Membership: |
|