Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01509832_m1
          See other GREM1 GE Assays ›
          Gene Symbol:
          Grem1
          Gene Name:
          gremlin 1
          Gene Aliases:
          Cktsf1b1, drm
          Chromosome Location:
          Chr.3: 105203309 - 105214989 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01509832_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grem1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019282.3 NP_062155.1 1 - 2 142 68
          GenBank mRNA Y10019.1 - 1 - 2 141 68

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_019282.3, Y10019.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grem1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          angiogenesis
          branching involved in ureteric bud morphogenesis
          cell migration involved in sprouting angiogenesis
          positive regulation of receptor internalization
          negative regulation of leukocyte chemotaxis
          positive regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation
          mesenchymal to epithelial transition involved in metanephros morphogenesis
          apoptotic process
          signal transduction
          activation of transmembrane receptor protein tyrosine kinase activity
          cell-cell signaling
          positive regulation of cell proliferation
          organ morphogenesis
          proximal/distal pattern formation
          regulation of epithelial to mesenchymal transition
          collagen fibril organization
          negative regulation of cell growth
          embryonic limb morphogenesis
          negative regulation of bone mineralization
          negative regulation of BMP signaling pathway
          negative regulation of chondrocyte differentiation
          regulation of stress-activated MAPK cascade
          negative regulation of osteoblast proliferation
          positive regulation of NF-kappaB import into nucleus
          negative regulation of apoptotic process
          endothelial cell migration
          positive regulation of angiogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of bone remodeling
          determination of dorsal identity
          positive regulation of NF-kappaB transcription factor activity
          regulation of focal adhesion assembly
          positive regulation of telomerase activity
          limb development
          negative regulation of pathway-restricted SMAD protein phosphorylation
          ureteric bud formation
          positive regulation of protein tyrosine kinase activity
          signal transduction by p53 class mediator
          negative regulation of monocyte chemotaxis
          negative regulation of canonical Wnt signaling pathway
          positive regulation of branching involved in ureteric bud morphogenesis
          negative regulation of branching involved in ureteric bud morphogenesis
          negative regulation of osteoclast proliferation
          positive regulation of peptidyl-tyrosine autophosphorylation
          negative regulation of bone trabecula formation
          negative regulation of bone mineralization involved in bone maturation
          positive regulation of receptor activity
          positive regulation of cardiac muscle cell differentiation
          cytokine activity
          protein binding
          transmembrane receptor protein tyrosine kinase activator activity
          vascular endothelial growth factor receptor 2 binding
          receptor agonist activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P550490 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P256301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754725 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66549 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P550490 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P256301 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P256301 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P550490 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P256301 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P550490 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P256301 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P550490 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78404 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10848282 - -
          Affymetrix Rat Gene 1.0 ST Array 10848285 - -
          Affymetrix Rat Gene 1.0 ST Array 10848286 - -
          Affymetrix Rat Gene 1.0 ST Array 10848284 - -
          Affymetrix Rat Gene 1.0 ST Array 10848283 - -
          Affymetrix Rat Gene 1.1 ST Array 10848281 - -
          Rat Genome 230 2.0 Array 1369113_PM_at - -
          Rat Genome 230 2.0 Array 1369113_at - -
          Affymetrix HT Rat Focus Array 1369113_PM_at - -
          Affymetrix HT Rat Focus Array 1369113_at - -
          Affymetrix Rat Clariom S Assay PSR0300029593.rn.1 TC0300003907.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300029594.rn.1 TC0300003907.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300029595.rn.1 TC0300003907.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300029596.rn.1 TC0300003907.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300029597.rn.1 TC0300003907.rn.1 RTA TC
          Rat Expression Set 230 1369113_PM_at - -
          Rat Expression Set 230 1369113_at - -
          Affymetrix Rat Gene 1.0 ST Array 10848281 - -
          Rat Genome U34 Set Y10019cds_at - -
          Rat Genome U34 Set Y10019cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21443176 - -
          ABI Rat Genome Survey Microarray 22197806 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355468 - -
          RatRef-12 Expression BeadChip ILMN_49412 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline