Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01510008_g1
          See other NDUFAB1 GE Assays ›
          Gene Symbol:
          Ndufab1
          Gene Name:
          NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 192044209 - 192057644 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01510008_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufab1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106294.1 NP_001099764.1 1 - 2 196 64
          XM_006230188.1 XP_006230250.1 1 - 2 228 64
          GenBank mRNA FQ212307.1 - 1 - 2 195 64
          FQ212475.1 - 1 - 2 233 64
          FQ216942.1 - 1 - 2 173 64

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein lipoylation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062201 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P075391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327873 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176699 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76593 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062201 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075391 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075391 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062201 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P075391 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062201 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P783113 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94717 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725479 - -
          Affymetrix Rat Gene 1.0 ST Array 10725477 - -
          Affymetrix Rat Gene 1.0 ST Array 10716463 - -
          Affymetrix Rat Gene 1.0 ST Array 10725478 - -
          Affymetrix Rat Gene 1.0 ST Array 10725475 - -
          Affymetrix Rat Gene 1.0 ST Array 10725476 - -
          Affymetrix Rat Gene 1.1 ST Array 10725474 - -
          Affymetrix Rat Gene 1.1 ST Array 10716461 - -
          Rat Genome 230 2.0 Array 1389964_PM_at - -
          Rat Genome 230 2.0 Array 1389964_at - -
          Affymetrix Rat Clariom S Assay PSR0100051496.rn.1 TC0100006558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051501.rn.1 TC0100006558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051504.rn.1 TC0100006558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051503.rn.1 TC0100006558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051505.rn.1 TC0100006558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051502.rn.1 TC0100006558.rn.1 RTA TC
          Rat Expression Set 230 1389964_PM_at - -
          Rat Expression Set 230 1389964_at - -
          Affymetrix Rat Gene 1.0 ST Array 10725474 - -
          Rat Genome U34 Set rc_AA891651_at - -
          Rat Genome U34 Set rc_AA891651_g_at - -
          Rat Genome U34 Set AA799336_at - -
          Rat Genome U34 Set rc_AA891651_at - -
          Rat Genome U34 Set AA799336_at - -
          Rat Genome U34 Set rc_AA891651_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21885206 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351396 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline