Search
Search
| Gene Symbol: Stx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012700.2 | NP_036832.1 | 6 - 7 | 518 | 105 | |
| XM_008759859.1 | XP_008758081.1 | 6 - 7 | 1055 | 105 | ||
| XM_008759860.1 | XP_008758082.1 | 6 - 7 | 456 | 105 | ||
| GenBank mRNA | M95735.1 | - | 6 - 7 | 677 | 105 | |
Set Membership: |
|