Search
Search
| Gene Symbol: Wnt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080401.1 | NP_536326.1 | 3 - 4 | 852 | 69 | |
| XM_006229718.2 | XP_006229780.1 | 3 - 4 | 704 | 69 | ||
| XM_006229719.2 | XP_006229781.1 | 4 - 5 | 1153 | 69 | ||
| XM_006229720.2 | XP_006229782.1 | 4 - 5 | 2075 | 69 | ||
| GenBank mRNA | AB073722.1 | - | 1 - 2 | 72 | 69 | |
Set Membership: |
|