Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01510618_g1
          See other TAF10 GE Assays ›
          Gene Symbol:
          Taf10
          Gene Name:
          TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 170585160 - 170586428 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01510618_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Taf10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134735.1 NP_001128207.1 3 - 4 470 58
          GenBank mRNA BC168203.1 - 3 - 4 470 7115651 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006299.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          liver development
          DNA-templated transcription, initiation
          regulation of transcription, DNA-templated
          transcription initiation from RNA polymerase II promoter
          apoptotic process
          biological_process
          regulation of gene expression
          histone deubiquitination
          multicellular organism growth
          cellular protein complex assembly
          histone H3 acetylation
          regulation of DNA binding
          protein homooligomerization
          hepatocyte differentiation
          molecular_function
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          histone acetyltransferase activity
          enzyme binding
          estrogen receptor binding
          RNA polymerase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P193221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968703 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P193221 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P193221 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P193221 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724528 - -
          Affymetrix Rat Gene 1.0 ST Array 10724525 - -
          Affymetrix Rat Gene 1.0 ST Array 10724526 - -
          Affymetrix Rat Gene 1.0 ST Array 10724527 - -
          Affymetrix Rat Gene 1.1 ST Array 10724524 - -
          Rat Genome 230 2.0 Array 1398963_PM_at - -
          Rat Genome 230 2.0 Array 1398963_at - -
          Affymetrix HT Rat Focus Array 1398963_PM_at - -
          Affymetrix HT Rat Focus Array 1398963_at - -
          Affymetrix Rat Clariom S Assay PSR0100049723.rn.1 TC0100006299.rn.1 RTA TC
          Rat Expression Set 230 1398963_at - -
          Rat Expression Set 230 1398963_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10724524 - -
          Rat Genome U34 Set rc_AA800015_at - -
          Rat Genome U34 Set rc_AA996740_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21174999 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364858 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline