Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01510868_m1
          See other EXNEF GE Assays ›
          Gene Symbol:
          Exnef
          Gene Name:
          exonuclease NEF-sp
          Gene Aliases:
          2610020h08rik, RGD1305412
          Chromosome Location:
          Chr.1: 189549960 - 189634033 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01510868_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Exnef
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001079706.1 NP_001073174.1 11 - 12 1224 123
          XM_006230111.2 XP_006230173.1 10 - 11 1154 123
          XM_006230121.2 XP_006230183.1 7 - 8 1106 123
          XM_008759751.1 XP_008757973.1 10 - 11 1160 123
          XM_008759752.1 XP_008757974.1 10 - 11 1445 123
          XM_008759753.1 XP_008757975.1 10 - 11 1446 123
          XM_008759755.1 XP_008757977.1 10 - 11 1445 123
          XM_008759756.1 XP_008757978.1 10 - 11 1445 123
          XM_008759757.1 XP_008757979.1 8 - 9 1205 123
          XM_008759759.1 XP_008757981.1 9 - 10 933 123
          XM_008759760.1 XP_008757982.1 10 - 11 1160 123
          GenBank mRNA BC128775.1 - 11 - 12 1224 7133646 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002485.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          exoribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleic acid phosphodiester bond hydrolysis
          nucleotide binding
          RNA binding
          exonuclease activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P250520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577128 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22660 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3372 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57387 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82056 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152482 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15288 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133028 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P250520 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P250520 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P250520 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P250520 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38888 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84037 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48505 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10710323 - -
          Affymetrix Rat Gene 1.0 ST Array 10710317 - -
          Affymetrix Rat Gene 1.0 ST Array 10710326 - -
          Affymetrix Rat Gene 1.0 ST Array 10710324 - -
          Affymetrix Rat Gene 1.0 ST Array 10710316 - -
          Affymetrix Rat Gene 1.0 ST Array 10710328 - -
          Affymetrix Rat Gene 1.0 ST Array 10710321 - -
          Affymetrix Rat Gene 1.0 ST Array 10710331 - -
          Affymetrix Rat Gene 1.0 ST Array 10710325 - -
          Affymetrix Rat Gene 1.0 ST Array 10710329 - -
          Affymetrix Rat Gene 1.0 ST Array 10710327 - -
          Affymetrix Rat Gene 1.0 ST Array 10710315 - -
          Affymetrix Rat Gene 1.0 ST Array 10710318 - -
          Affymetrix Rat Gene 1.0 ST Array 10710320 - -
          Affymetrix Rat Gene 1.0 ST Array 10710322 - -
          Affymetrix Rat Gene 1.0 ST Array 10710332 - -
          Affymetrix Rat Gene 1.0 ST Array 10710330 - -
          Affymetrix Rat Gene 1.0 ST Array 10710319 - -
          Affymetrix Rat Gene 1.0 ST Array 10710314 - -
          Affymetrix Rat Gene 1.1 ST Array 10710313 - -
          Affymetrix Rat Clariom S Assay PSR0100018913.rn.1 TC0100002485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018921.rn.1 TC0100002485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018927.rn.1 TC0100002485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018914.rn.1 TC0100002485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018917.rn.1 TC0100002485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018919.rn.1 TC0100002485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018922.rn.1 TC0100002485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018926.rn.1 TC0100002485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018915.rn.1 TC0100002485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018918.rn.1 TC0100002485.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018920.rn.1 TC0100002485.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10710313 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20926397 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361409 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline