Search
Search
| Gene Symbol: Relt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108495.1 | NP_001101965.1 | 5 - 6 | 652 | 106 | |
| XM_006229775.2 | XP_006229837.1 | 6 - 7 | 820 | 106 | ||
| XM_006229776.2 | XP_006229838.1 | 6 - 7 | 3084 | 106 | ||
| XM_006229777.2 | XP_006229839.1 | 6 - 7 | 771 | 106 | ||
| XM_006229778.2 | XP_006229840.1 | 6 - 7 | 5172 | 106 | ||
| XM_006229780.2 | XP_006229842.1 | 6 - 7 | 820 | 106 | ||
| XM_008759763.1 | XP_008757985.1 | 6 - 7 | 5500 | 106 | ||
| XM_008759764.1 | XP_008757986.1 | 6 - 7 | 5425 | 106 | ||
| XM_008759765.1 | XP_008757987.1 | 6 - 7 | 5380 | 106 | ||
Set Membership: |
|