Search
Search
| Gene Symbol: Taf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108197.1 | NP_001101667.1 | 3 - 4 | 314 | 63 | |
| XM_008766860.1 | XP_008765082.1 | 3 - 4 | 315 | 63 | ||
| XM_008766861.1 | XP_008765083.1 | 3 - 4 | 315 | 63 | ||
| XM_008766862.1 | XP_008765084.1 | 3 - 4 | 315 | 63 | ||
| GenBank mRNA | FQ211123.1 | - | 3 - 4 | 311 | 63 | |
Set Membership: |
|