Search
Search
| Gene Symbol: Api5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127379.1 | NP_001120851.1 | 7 - 8 | 926 | 87 | |
| XM_006234586.1 | XP_006234648.1 | 7 - 8 | 1019 | 87 | ||
| GenBank mRNA | BC162003.1 | - | 7 - 8 | 926 | 9089120 | 87 |
| DQ480749.1 | - | 7 - 8 | 1003 | 87 | ||
Set Membership: |
|