Search
Search
| Gene Symbol: Osbpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107735.1 | NP_001101205.1 | 9 - 10 | 1258 | 60 | |
| XM_008761900.1 | XP_008760122.1 | 6 - 7 | 677 | 60 | ||
| XM_008761901.1 | XP_008760123.1 | 6 - 7 | 677 | 60 | ||
| XM_008761902.1 | XP_008760124.1 | 6 - 7 | 677 | 60 | ||
| XM_008761903.1 | XP_008760125.1 | 6 - 7 | 677 | 60 | ||
| XM_008761904.1 | XP_008760126.1 | 6 - 7 | 677 | 60 | ||
| XM_008761905.1 | XP_008760127.1 | 6 - 7 | 677 | 60 | ||
| XM_008761906.1 | XP_008760128.1 | 6 - 7 | 677 | 60 | ||
| XM_008761907.1 | XP_008760129.1 | 6 - 7 | 677 | 60 | ||
Set Membership: |
|