Search
Search
| Gene Symbol: Spi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005892.2 | NP_001005892.1 | 2 - 3 | 296 | 67 | |
| XM_006234532.2 | XP_006234594.1 | 2 - 3 | 1175 | 67 | ||
| GenBank mRNA | AB154364.1 | - | 2 - 3 | 159 | 67 | |
| BC091185.1 | - | 2 - 3 | 296 | 7366435 | 67 | |
Set Membership: |
|