Search
Search
| Gene Symbol: Scrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013162.1 | NP_001013180.1 | 1 - 2 | 49 | 84 | |
| XM_006234358.2 | XP_006234420.1 | 1 - 2 | 86 | 84 | ||
| XM_006234359.2 | XP_006234421.1 | 1 - 2 | 88 | 84 | ||
| XM_006234360.2 | XP_006234422.1 | 1 - 2 | 88 | 84 | ||
| GenBank mRNA | BC088234.1 | - | 1 - 2 | 49 | 7324882 | 84 |
Set Membership: |
|