Search
Search

Hnrnpa3| Gene Symbol: Hnrnpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198132.3 | NP_937765.1 | 10 - 11 | 1238 | 98 | |
| XM_008761912.1 | XP_008760134.1 | 10 - 11 | 1187 | 98 | ||
| GenBank mRNA | BC081878.1 | - | 10 - 11 | 1178 | 7108128 | 98 |
| FQ214617.1 | - | 10 - 11 | 1134 | 98 | ||
Set Membership: |
|