Search
Search
| Gene Symbol: Nusap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107762.1 | NP_001101232.1 | 1 - 2 | 356 | 82 | |
| XM_006234768.2 | XP_006234830.1 | 1 - 2 | 187 | 82 | ||
| XM_006234769.2 | XP_006234831.1 | 1 - 2 | 187 | 82 | ||
| XM_006234770.2 | XP_006234832.1 | 1 - 2 | 187 | 82 | ||
| XM_008762101.1 | XP_008760323.1 | 1 - 2 | 187 | 82 | ||
| XM_008762102.1 | XP_008760324.1 | 1 - 2 | 187 | 82 | ||
| GenBank mRNA | BC161947.1 | - | 1 - 2 | 132 | 9025380 | 82 |
Set Membership: |
|