Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01515209_g1
          See other NUSAP1 GE Assays ›
          Gene Symbol:
          Nusap1
          Gene Name:
          nucleolar and spindle associated protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 111422267 - 111452600 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01515209_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nusap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107762.1 NP_001101232.1 9 - 10 1303 74
          XM_006234768.2 XP_006234830.1 10 - 11 1254 74
          XM_006234769.2 XP_006234831.1 10 - 11 1251 74
          XM_006234770.2 XP_006234832.1 9 - 10 1107 74
          XM_008762101.1 XP_008760323.1 9 - 10 1164 74
          XM_008762102.1 XP_008760324.1 9 - 10 1161 74
          GenBank mRNA BC161947.1 - 10 - 11 1199 9025380 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic sister chromatid segregation
          mitotic cytokinesis
          mitotic chromosome condensation
          establishment of mitotic spindle localization
          positive regulation of mitotic nuclear division
          microtubule binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P818287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25591 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39220 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34980 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P818287 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P818287 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P818287 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P511128 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64259 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5106 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10838845 - -
          Affymetrix Rat Gene 1.0 ST Array 10838853 - -
          Affymetrix Rat Gene 1.0 ST Array 10838850 - -
          Affymetrix Rat Gene 1.0 ST Array 10838846 - -
          Affymetrix Rat Gene 1.0 ST Array 10838849 - -
          Affymetrix Rat Gene 1.0 ST Array 10838848 - -
          Affymetrix Rat Gene 1.0 ST Array 10838851 - -
          Affymetrix Rat Gene 1.0 ST Array 10838852 - -
          Affymetrix Rat Gene 1.0 ST Array 10838844 - -
          Affymetrix Rat Gene 1.0 ST Array 10838847 - -
          Affymetrix Rat Gene 1.1 ST Array 10838843 - -
          Rat Genome 230 2.0 Array 1392732_PM_at - -
          Rat Genome 230 2.0 Array 1392732_at - -
          Rat Genome 230 2.0 Array 1384280_PM_at - -
          Rat Genome 230 2.0 Array 1384280_at - -
          Affymetrix HT Rat Focus Array 1392732_PM_at - -
          Affymetrix HT Rat Focus Array 1392732_at - -
          Affymetrix HT Rat Focus Array 1384280_PM_at - -
          Affymetrix HT Rat Focus Array 1384280_at - -
          Affymetrix Rat Clariom S Assay PSR0300010282.rn.1 TC0300001518.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300010283.rn.1 TC0300001518.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300010287.rn.1 TC0300001518.rn.1 RTA TC
          Rat Expression Set 230 1384280_at - -
          Rat Expression Set 230 1392732_PM_at - -
          Rat Expression Set 230 1384280_PM_at - -
          Rat Expression Set 230 1392732_at - -
          Affymetrix Rat Gene 1.0 ST Array 10838843 - -
          Rat Genome U34 Set rc_AA964152_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21464514 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351744 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline