Search
Search

Creb3l1| Gene Symbol: Creb3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005562.1 | NP_001005562.1 | 3 - 4 | 908 | 68 | |
| XM_006234584.1 | XP_006234646.1 | 3 - 4 | 964 | 68 | ||
| GenBank mRNA | BC081951.1 | - | 3 - 4 | 908 | 7122657 | 68 |
Set Membership: |
|