Search
Search
| Gene Symbol: Txlna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127633.1 | NP_001121105.1 | 5 - 6 | 889 | 83 | |
| XM_006238972.2 | XP_006239034.1 | 5 - 6 | 896 | 83 | ||
| GenBank mRNA | BC166486.1 | - | 5 - 6 | 889 | 7099983 | 83 |
Set Membership: |
|